Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017643.2 | NP_060113.2 | 9 | 1156 | ||
XM_005257467.4 | XP_005257524.1 | 8 | 1101 | |||
XM_005257468.4 | XP_005257525.1 | 8 | 1094 | |||
XM_011524922.2 | XP_011523224.1 | 8 | 1103 | |||
XM_011524923.2 | XP_011523225.1 | 8 | 1100 | |||
XM_011524924.2 | XP_011523226.1 | 8 | 1096 | |||
XM_011524925.1 | XP_011523227.1 | 9 | 1217 | |||
XM_011524926.2 | XP_011523228.1 | 9 | 1321 | |||
XM_011524928.2 | XP_011523230.1 | 8 | 1258 | |||
XM_011524929.2 | XP_011523231.1 | 9 | 1172 | |||
XM_011524930.1 | XP_011523232.1 | 8 | 894 | |||
XM_011524931.2 | XP_011523233.1 | 7 | 728 | |||
XM_011524933.2 | XP_011523235.1 | 10 | 1278 | |||
XM_011524934.2 | XP_011523236.1 | 8 | 1102 | |||
XM_011524935.2 | XP_011523237.1 | 8 | 1095 | |||
XM_011524936.1 | XP_011523238.1 | 8 | 894 | |||
XM_017024782.1 | XP_016880271.1 | 9 | 1468 | |||
GenBank mRNA | AK000062.1 | - | 5 | 488 | ||
AK124061.1 | - | 8 | 894 | |||
BC034364.1 | - | 9 | 1127 | |||
BC101736.1 | - | 4 | 417 | 8069242 |