Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270890.1 | NP_001257819.1 | 16 | 2944 | ||
NM_017645.4 | NP_060115.3 | 16 | 3049 | |||
XM_011517935.2 | XP_011516237.1 | 5 | 1897 | |||
XM_017014830.1 | XP_016870319.1 | 12 | 2200 | |||
GenBank mRNA | AB046794.1 | - | 9 | 1905 | ||
AF533709.1 | - | 16 | 3231 | |||
AF537091.1 | - | 16 | 3011 | |||
AK000067.1 | - | 2 | 1137 | |||
AK022964.1 | - | 13 | 2365 | |||
AK023068.1 | - | 9 | 1968 | |||
AK056458.1 | - | 16 | 3011 | |||
AK225209.1 | - | 2 | 1137 | |||
AK301852.1 | - | 9 | 1804 | |||
AL832495.1 | - | 16 | 3009 | |||
BC010632.1 | - | 2 | 1006 | |||
BC065935.1 | - | 14 | 2549 | |||
BC111041.1 | - | 12 | 2325 | 5529130 | ||
BC113981.1 | - | 16 | 2846 | 40082296 | ||
BC114492.1 | - | 16 | 2846 | 40082291 |