Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF562 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130031.1 | NP_001123503.1 | 6 | 1364 | ||
NM_001130032.1 | NP_001123504.1 | 6 | 1311 | |||
NM_001300885.1 | NP_001287814.1 | 6 | 1361 | |||
NM_017656.3 | NP_060126.2 | 4 | 1148 | |||
XM_005259941.3 | XP_005259998.1 | 6 | 1731 | |||
XM_005259944.3 | XP_005260001.1 | 5 | 1467 | |||
XM_005259951.3 | XP_005260008.1 | 5 | 1426 | |||
XM_005259952.3 | XP_005260009.1 | 5 | 1448 | |||
XM_011528100.2 | XP_011526402.1 | 4 | 1310 | |||
XM_011528102.2 | XP_011526404.1 | 3 | 1234 | |||
XM_017026898.1 | XP_016882387.1 | 6 | 1728 | |||
XM_017026899.1 | XP_016882388.1 | 5 | 1500 | |||
GenBank mRNA | AK000086.1 | - | 4 | 1106 | ||
AK297446.1 | - | 6 | 1345 | |||
AK300115.1 | - | 5 | 1214 | |||
AK303038.1 | - | 5 | 1262 | |||
AK304370.1 | - | 4 | 1120 | |||
AY346375.1 | - | 5 | 1148 | |||
BC109062.2 | - | 5 | 1254 | 40033739 |