Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QPCTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017659.3 | NP_060129.2 | 3 | 727 | ||
XM_011527048.2 | XP_011525350.1 | 3 | 808 | |||
XM_017026900.1 | XP_016882389.1 | 3 | 806 | |||
GenBank mRNA | AK000091.1 | - | 3 | 536 | ||
AK172764.1 | - | 3 | 535 | |||
AK222636.1 | - | 3 | 520 |