Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143979.1 | NP_001137451.1 | 7 | 1492 | ||
NM_017668.2 | NP_060138.1 | 6 | 778 | |||
XM_005255396.4 | XP_005255453.1 | 6 | 771 | |||
XM_006720897.3 | XP_006720960.1 | 6 | 1338 | |||
XM_006720900.3 | XP_006720963.1 | 6 | 807 | |||
XM_011522553.2 | XP_011520855.1 | 7 | 1492 | |||
XM_017023349.1 | XP_016878838.1 | 6 | 771 | |||
XM_017023350.1 | XP_016878839.1 | 6 | 761 | |||
XM_017023351.1 | XP_016878840.1 | 6 | 771 | |||
XM_017023352.1 | XP_016878841.1 | 6 | 771 | |||
XM_017023353.1 | XP_016878842.1 | 6 | 771 | |||
XM_017023354.1 | XP_016878843.1 | 6 | 771 | |||
XM_017023355.1 | XP_016878844.1 | 6 | 771 | |||
XM_017023356.1 | XP_016878845.1 | 6 | 807 | |||
XM_017023357.1 | XP_016878846.1 | 4 | 525 | |||
GenBank mRNA | AF124431.1 | - | 5 | 687 | ||
AK000108.1 | - | 6 | 757 | |||
AK025815.1 | - | 6 | 757 | |||
AK127927.1 | - | 3 | 1145 | |||
BC001421.2 | - | 6 | 771 | 3140369 | ||
KJ534887.1 | - | 5 | 666 | |||
KJ534967.1 | - | 5 | 666 | |||
KJ534991.1 | - | 5 | 666 | |||
KJ535028.1 | - | 4 | 517 |