Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317954.1 | NP_001304883.1 | 4 | 670 | ||
NM_017676.2 | NP_060146.2 | 5 | 864 | |||
XM_005272028.3 | XP_005272085.1 | 5 | 871 | |||
XM_017009599.1 | XP_016865088.1 | 5 | 2299 | |||
GenBank mRNA | AK000132.1 | - | 5 | 862 | ||
AK225572.1 | - | 5 | 861 | |||
AK295796.1 | - | 5 | 852 | |||
BC151135.1 | - | 4 | 804 | 9057035 | ||
BC157835.1 | - | 4 | 804 | 9057054 | ||
CR627424.1 | - | 4 | 641 |