Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017694.3 | NP_060164.3 | 3 | 872 | ||
XM_005246654.2 | XP_005246711.1 | 2 | 750 | |||
XM_005246656.3 | XP_005246713.1 | 2 | 750 | |||
XM_011511368.1 | XP_011509670.1 | 4 | 938 | |||
XM_017004392.1 | XP_016859881.1 | 3 | 872 | |||
GenBank mRNA | AB472299.1 | - | 1 | 548 | ||
AK098085.1 | - | 1 | 283 | |||
AK127176.1 | - | 3 | 872 |