Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYTL2 SIRNA Assays ›
Gene Symbol:
SYTL2
Gene Name:
synaptotagmin like 2
Gene Aliases:
CHR11SYT, EXO4, PPP1R151, SGA72M, SLP2, SLP2A
Chromosome Location:
Chr.11: 85694221 - 85811182 on Build GRCh38
UniGene ID:
Hs.369520
Species:
Human
siRNA ID
HSS123356
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYTL2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001162951.2
NP_001156423.1
17
2986
NM_001162952.2
NP_001156424.1
9
1516
NM_001162953.2
NP_001156425.1
17
2989
NM_001289608.1
NP_001276537.1
18
2795
NM_001289609.1
NP_001276538.1
10
1629
NM_001289610.1
NP_001276539.1
10
1200
NM_032943.4
NP_116561.1
18
3165
NM_206929.3
NP_996812.1
11
1320
NM_206930.3
NP_996813.1
8
1493
XM_005274057.1
XP_005274114.1
17
6625
XM_005274060.1
XP_005274117.1
18
2809
XM_005274061.1
XP_005274118.1
18
2806
XM_005274062.2
XP_005274119.1
19
3230
XM_005274063.1
XP_005274120.1
17
2689
XM_005274066.1
XP_005274123.1
16
2638
XM_005274067.4
XP_005274124.1
10
2932
XM_011545105.1
XP_011543407.1
18
6673
XM_011545106.1
XP_011543408.1
18
6670
XM_011545107.2
XP_011543409.1
19
7094
XM_011545108.1
XP_011543410.1
17
6553
XM_011545109.2
XP_011543411.1
18
6655
XM_011545110.1
XP_011543412.1
17
6502
XM_011545114.2
XP_011543416.1
9
2838
XM_011545115.1
XP_011543417.1
7
932
XM_011545116.2
XP_011543418.1
8
2717
XM_017017933.1
XP_016873422.1
17
6622
XM_017017934.1
XP_016873423.1
19
7091
XM_017017935.1
XP_016873424.1
15
6469
XM_017017936.1
XP_016873425.1
15
6167
XM_017017937.1
XP_016873426.1
18
2710
XM_017017938.1
XP_016873427.1
17
2668
XM_017017939.1
XP_016873428.1
11
2976
XM_017017940.1
XP_016873429.1
10
2928
GenBank mRNA
AB046817.1
-
18
3176
AK024872.1
-
8
1657
AK074737.1
-
11
1277
AK092121.1
-
8
1311
AK124754.1
-
11
2901
AK127504.1
-
12
3863
AK222707.1
-
8
1657
AK222710.1
-
8
1657
AK296224.1
-
18
2777
AK296740.1
-
15
2725
AK298604.1
-
10
1118
AL834422.1
-
5
555
AY386362.1
-
11
5180
BC015540.2
-
8
987
BC068495.1
-
10
1352
BC110315.1
-
9
1045
BC136450.1
-
17
2921
9020439
BC144114.1
-
17
2921
9052634
BC144115.1
-
17
2918
9052635
BX647671.1
-
12
3616
BX648068.1
-
11
3062
EU035829.1
-
17
2642
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
exocytosis
vesicle docking involved in exocytosis
vesicle fusion
negative regulation of phosphatase activity
vesicle-mediated transport
regulation of calcium ion-dependent exocytosis
calcium ion-regulated exocytosis of neurotransmitter
positive regulation of mucus secretion
protein localization to plasma membrane
phosphatidylserine binding
calcium ion binding
protein binding
calcium-dependent phospholipid binding
phosphatidylinositol-4,5-bisphosphate binding
Rab GTPase binding
phosphatase binding
syntaxin binding
clathrin binding
neurexin family protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline