Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIDT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308350.1 | NP_001295279.1 | 3 | 1074 | ||
NM_001322294.1 | NP_001309223.1 | 3 | 1074 | |||
NM_001322295.1 | NP_001309224.1 | 3 | 1074 | |||
NM_001322296.1 | NP_001309225.1 | 3 | 1074 | |||
NM_001322297.1 | NP_001309226.1 | 2 | 952 | |||
NM_001322298.1 | NP_001309227.1 | 3 | 1074 | |||
NM_001322299.1 | NP_001309228.1 | 3 | 1074 | |||
NM_001322300.1 | NP_001309229.1 | 3 | 1074 | |||
NM_017699.2 | NP_060169.2 | 3 | 1074 | |||
NR_136280.1 | 4 | 1154 | ||||
NR_136281.1 | 3 | 1074 | ||||
NR_136282.1 | 3 | 1074 | ||||
NR_136283.1 | 3 | 1074 | ||||
NR_136284.1 | 3 | 1074 | ||||
NR_136285.1 | 3 | 1074 | ||||
XM_011512939.2 | XP_011511241.1 | 3 | 1034 | |||
XM_011512941.2 | XP_011511243.1 | 3 | 1034 | |||
XM_017006664.1 | XP_016862153.1 | 3 | 1034 | |||
XM_017006665.1 | XP_016862154.1 | 2 | 1549 | |||
GenBank mRNA | BC117222.1 | - | 3 | 462 | 40125773 | |
BC143776.1 | - | 3 | 462 | |||
BC143778.1 | - | 3 | 462 | |||
DA217300.1 | - | 3 | 418 |