Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIDT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308350.1 | NP_001295279.1 | 5 | 1264 | ||
NM_001322294.1 | NP_001309223.1 | 5 | 1264 | |||
NM_001322295.1 | NP_001309224.1 | 5 | 1264 | |||
NM_001322297.1 | NP_001309226.1 | 4 | 1142 | |||
NM_001322298.1 | NP_001309227.1 | 5 | 1264 | |||
NM_001322299.1 | NP_001309228.1 | 5 | 1264 | |||
NM_017699.2 | NP_060169.2 | 5 | 1264 | |||
NR_136280.1 | 6 | 1344 | ||||
NR_136281.1 | 5 | 1264 | ||||
NR_136282.1 | 5 | 1264 | ||||
NR_136283.1 | 5 | 1264 | ||||
XM_011512939.2 | XP_011511241.1 | 5 | 1224 | |||
XM_011512941.2 | XP_011511243.1 | 5 | 1224 | |||
XM_017006664.1 | XP_016862153.1 | 4 | 1178 | |||
XM_017006665.1 | XP_016862154.1 | 4 | 1739 | |||
XM_017006666.1 | XP_016862155.1 | 2 | 132 | |||
GenBank mRNA | BC117222.1 | - | 5 | 652 | 40125773 | |
BC143776.1 | - | 5 | 652 | |||
BC143778.1 | - | 5 | 652 |