Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316937.1 | NP_001303866.1 | 2, 3 | 426 | ||
NM_001316940.1 | NP_001303869.1 | 2, 3 | 348 | |||
NM_017703.2 | NP_060173.1 | 2, 3 | 452 | |||
GenBank mRNA | AK000195.1 | - | 2, 3 | 404 | ||
AK093760.1 | - | 2, 3 | 399 | |||
BC001586.2 | - | 2, 3 | 356 | 2966439 | ||
BP367273.1 | - | 2, 3 | 321 | |||
BX647073.1 | - | 1, 2 | 289 | |||
DB127388.1 | - | 2, 3 | 426 |