Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017706.4 | NP_060176.2 | 4 | 510 | ||
XM_005268469.2 | XP_005268526.1 | 4 | 510 | |||
XM_017009600.1 | XP_016865089.1 | 5 | 297 | |||
GenBank mRNA | AK056861.1 | - | 3 | 521 | ||
AK122862.1 | - | 2 | 1945 | |||
BC002482.2 | - | 3 | 216 | |||
BC041687.1 | - | 4 | 392 | |||
BQ069026.1 | - | 4 | 481 | |||
DB210296.1 | - | 4 | 510 |