Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM46C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017709.3 | NP_060179.2 | 2 | 324 | ||
GenBank mRNA | AK000209.1 | - | 2 | 259 | ||
BC036516.2 | - | 2 | 253 | 5267660 | ||
BC131726.1 | - | 2 | 237 | 40123993 | ||
CD300120.1 | - | 2 | 350 |