Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171191.1 | NP_001164662.1 | 2 | 417 | ||
NM_001171192.1 | NP_001164663.1 | 4 | 635 | |||
NM_001171193.1 | NP_001164664.1 | 3 | 521 | |||
NM_017711.3 | NP_060181.2 | 4 | 635 | |||
XM_017029614.1 | XP_016885103.1 | 3 | 521 | |||
GenBank mRNA | AB048363.1 | - | 4 | 525 | ||
AK000214.1 | - | 4 | 347 | |||
AK299255.1 | - | 2 | 245 | |||
AK301025.1 | - | 4 | 459 | |||
AK307778.1 | - | 4 | 336 | |||
AK316035.1 | - | 3 | 253 | |||
AY358986.1 | - | 4 | 330 | |||
BC032009.1 | - | 4 | 332 | 4747934 |