Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGPEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300927.1 | NP_001287856.1 | 1, 2 | 128 | ||
NM_001308366.1 | NP_001295295.1 | 1, 2 | 128 | |||
NM_017712.3 | NP_060182.1 | 1, 2 | 128 | |||
XM_006722783.3 | XP_006722846.1 | 1, 2 | 142 | |||
XM_011528111.2 | XP_011526413.1 | 1, 2 | 128 | |||
XM_011528112.1 | XP_011526414.1 | 1, 2 | 128 | |||
GenBank mRNA | AJ278828.1 | - | 1, 2 | 75 | ||
AK000215.1 | - | 1, 2 | 75 | |||
AK289968.1 | - | 1, 2 | 128 | |||
BC028063.1 | - | 1, 2 | 101 | |||
BC042138.1 | - | 1, 2 | 104 | 5935845 | ||
DA315740.1 | - | 1, 2 | 128 | |||
DA738481.1 | - | 1, 2 | 117 |
Gene Symbol: FKBP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK026952.1 | - | 72 |