Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031703.2 | NP_001026873.2 | 1 | 134 | ||
XM_005265241.3 | XP_005265298.1 | 1 | 131 | |||
XM_011533851.2 | XP_011532153.1 | 1 | 136 | |||
XM_017006669.1 | XP_016862158.1 | 1 | 136 | |||
GenBank mRNA | AK000218.1 | - | 1 | 119 | ||
AK225721.1 | - | 1 | 116 | |||
AK298202.1 | - | 1 | 141 | |||
BC000623.2 | - | 1 | 38 | 3346325 | ||
BC014204.2 | - | 1 | 38 | 4620013 | ||
DQ787088.1 | - | 1 | 15 |