Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083590.1 | NP_001077059.1 | 14 | 1690 | ||
NM_017727.4 | NP_060197.4 | 15 | 1825 | |||
XM_005264381.3 | XP_005264438.1 | 16 | 1990 | |||
XM_005264382.3 | XP_005264439.1 | 16 | 1876 | |||
XM_005264383.3 | XP_005264440.1 | 8 | 873 | |||
GenBank mRNA | AF289555.1 | - | 9 | 861 | ||
AK000261.1 | - | 10 | 942 | |||
AK074981.1 | - | 15 | 1753 | |||
AK097001.1 | - | 1 | 759 | |||
AK097308.1 | - | 14 | 1620 | |||
AK296575.1 | - | 8 | 925 | |||
AK315359.1 | - | 13 | 1289 | |||
AL833725.1 | - | 15 | 1685 | |||
AL834157.1 | - | 10 | 930 | |||
BC068479.1 | - | 15 | 1777 | 4814193 |