Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIGG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127178.2 | NP_001120650.1 | 13 | 2957 | ||
NM_001289051.1 | NP_001275980.1 | 13 | 2766 | |||
NM_001289052.1 | NP_001275981.1 | 11 | 2558 | |||
NM_017733.4 | NP_060203.3 | 13 | 2933 | |||
NR_110293.1 | 13 | 2911 | ||||
XM_005272284.2 | XP_005272341.1 | 13 | 3191 | |||
XM_011513491.1 | XP_011511793.1 | 14 | 2987 | |||
XM_011513492.1 | XP_011511794.1 | 8 | 1841 | |||
XM_017008321.1 | XP_016863810.1 | 13 | 3167 | |||
XM_017008322.1 | XP_016863811.1 | 12 | 3049 | |||
XM_017008323.1 | XP_016863812.1 | 14 | 3305 | |||
XM_017008324.1 | XP_016863813.1 | 13 | 3145 | |||
XM_017008325.1 | XP_016863814.1 | 14 | 2996 | |||
GenBank mRNA | AB162713.1 | - | 13 | 2927 | ||
AK000272.1 | - | 8 | 1723 | |||
AK027465.1 | - | 7 | 1676 | |||
AK074715.1 | - | 13 | 2729 | |||
AK074815.1 | - | 13 | 2912 | |||
AL137741.1 | - | 2 | 3228 | |||
AL157494.1 | - | 2 | 1954 | |||
AL162084.1 | - | 2 | 436 | |||
AW451782.1 | - | 1 | 306 | |||
BC000937.2 | - | 5 | 1103 | |||
BC001249.1 | - | 9 | 1959 | |||
BC110878.1 | - | 11 | 2500 | 5750347 | ||
BC144576.1 | - | 13 | 2868 |