Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PALMD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017734.4 | NP_060204.1 | 7 | 1662 | ||
GenBank mRNA | AF262379.1 | - | 7 | 1593 | ||
AJ312214.1 | - | 7 | 1504 | |||
AK000278.1 | - | 7 | 1424 | |||
AK024654.1 | - | 2 | 725 | |||
AL390969.1 | - | 1 | 466 | |||
AL390970.1 | - | 1 | 691 | |||
AL390972.1 | - | 7 | 1101 | |||
AL390973.1 | - | 7 | 1388 | |||
BC012570.1 | - | 7 | 1477 | 4245261 | ||
BX648213.1 | - | 4 | 4377 | |||
KU178657.1 | - | 7 | 1219 |