Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNBP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024948.2 | NP_001020119.1 | 13 | 1608 | ||
NM_001164473.2 | NP_001157945.1 | 15 | 1782 | |||
NM_017737.4 | NP_060207.2 | 13 | 1608 | |||
XM_011541625.2 | XP_011539927.1 | 13 | 1672 | |||
XM_017001533.1 | XP_016857022.1 | 14 | 1803 | |||
GenBank mRNA | AK000282.1 | - | 7 | 860 | ||
AK296767.1 | - | 4 | 449 | |||
AK299760.1 | - | 10 | 1168 | |||
AK299943.1 | - | 9 | 1078 | |||
AK315981.1 | - | 7 | 792 | |||
AY514449.1 | - | 13 | 1457 | |||
BC062477.1 | - | 8 | 1059 | |||
BC074891.2 | - | 7 | 838 | |||
BC074892.2 | - | 7 | 838 |