Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161584.1 | NP_001155056.1 | 3 | 670 | ||
NM_017746.3 | NP_060216.2 | 3 | 726 | |||
XM_011518798.1 | XP_011517100.1 | 3 | 688 | |||
GenBank mRNA | AB060968.1 | - | 3 | 642 | ||
AK000294.1 | - | 3 | 642 | |||
AK302619.1 | - | 3 | 673 | |||
AK304869.1 | - | 2 | 443 | |||
AY598337.1 | - | 2 | 511 | |||
BC030652.2 | - | 3 | 562 | 5268105 |