Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAPG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281932.1 | NP_001268861.1 | 22 | 2817 | ||
NM_001281933.1 | NP_001268862.1 | 21 | 2764 | |||
NM_017760.6 | NP_060230.5 | 21 | 2764 | |||
NR_104054.1 | 24 | 2975 | ||||
XM_005249547.2 | XP_005249604.1 | 21 | 2762 | |||
XM_011516356.2 | XP_011514658.1 | 21 | 2762 | |||
XM_011516357.2 | XP_011514659.1 | 22 | 2790 | |||
XM_011516358.2 | XP_011514660.1 | 21 | 2762 | |||
XM_011516362.1 | XP_011514664.1 | 9 | 1241 | |||
XM_017012374.1 | XP_016867863.1 | 21 | 2762 | |||
XM_017012375.1 | XP_016867864.1 | 20 | 2588 | |||
XM_017012376.1 | XP_016867865.1 | 20 | 2588 | |||
XM_017012377.1 | XP_016867866.1 | 11 | 1520 | |||
XM_017012378.1 | XP_016867867.1 | 18 | 2167 | |||
GenBank mRNA | AK000318.1 | - | 16 | 2022 | ||
AK022744.1 | - | 11 | 1564 | |||
AK023347.1 | - | 10 | 1318 | |||
AK092008.1 | - | 22 | 2752 | |||
AK295057.1 | - | 7 | 983 | |||
AK301901.1 | - | 9 | 1242 | |||
AK315699.1 | - | 18 | 2461 | |||
BC020560.1 | - | 16 | 2088 | 3458938 | ||
BC043404.1 | - | 21 | 2705 | 6045248 | ||
BX537845.1 | - | 21 | 2731 |