Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271420.1 | NP_001258349.1 | 9 | 1493 | ||
NM_017775.3 | NP_060245.3 | 9 | 1356 | |||
XM_017024801.1 | XP_016880290.1 | 9 | 1269 | |||
XM_017024802.1 | XP_016880291.1 | 9 | 1269 | |||
GenBank mRNA | AK000350.1 | - | 7 | 572 | ||
AK025958.1 | - | 3 | 242 | |||
AK055780.1 | - | 9 | 1049 | |||
AK056878.1 | - | 9 | 1268 | |||
AK094819.1 | - | 2 | 1550 | |||
AK297783.1 | - | 9 | 912 | |||
AL832412.1 | - | 2 | 610 | |||
BC011698.2 | - | 9 | 895 | |||
BC073796.1 | - | 9 | 899 | |||
BC105128.1 | - | 9 | 1356 | 8144131 | ||
BC112107.1 | - | 9 | 1356 | 8327575 |