Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165927.1 | NP_001159399.1 | 11 | 1063 | ||
NM_001321268.1 | NP_001308197.1 | 10 | 976 | |||
NM_001321269.1 | NP_001308198.1 | 11 | 1074 | |||
NM_017777.3 | NP_060247.2 | 11 | 1074 | |||
XM_005257485.3 | XP_005257542.1 | 11 | 1055 | |||
XM_006721965.2 | XP_006722028.1 | 10 | 908 | |||
XM_011524957.2 | XP_011523259.1 | 11 | 1054 | |||
XM_011524958.2 | XP_011523260.1 | 11 | 1054 | |||
XM_011524959.2 | XP_011523261.1 | 11 | 1053 | |||
XM_011524960.2 | XP_011523262.1 | 11 | 1053 | |||
XM_017024803.1 | XP_016880292.1 | 11 | 1022 | |||
XM_017024804.1 | XP_016880293.1 | 11 | 1022 | |||
XM_017024805.1 | XP_016880294.1 | 11 | 1088 | |||
GenBank mRNA | AK000352.1 | - | 8 | 744 | ||
AK301020.1 | - | 10 | 907 | |||
AK310815.1 | - | 11 | 1063 | |||
BC010061.2 | - | 11 | 1009 | 3952532 | ||
CR457229.1 | - | 7 | 570 | |||
DQ185029.1 | - | 11 | 1074 | |||
KU178661.1 | - | 7 | 570 | |||
KU178662.1 | - | 7 | 570 | |||
KU178663.1 | - | 7 | 570 |