Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPDL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017785.4 | NP_060255.3 | 6 | 1024 | ||
XM_011534585.1 | XP_011532887.1 | 6 | 970 | |||
XM_011534586.2 | XP_011532888.1 | 6 | 906 | |||
XM_011534587.2 | XP_011532889.1 | 7 | 1171 | |||
XM_011534588.2 | XP_011532890.1 | 7 | 1109 | |||
XM_011534589.2 | XP_011532891.1 | 6 | 919 | |||
XM_011534590.2 | XP_011532892.1 | 5 | 660 | |||
XM_017009601.1 | XP_016865090.1 | 6 | 855 | |||
XM_017009602.1 | XP_016865091.1 | 5 | 660 | |||
GenBank mRNA | AK000371.1 | - | 5 | 750 | ||
AK074227.1 | - | 3 | 429 | |||
AK304623.1 | - | 5 | 688 | |||
AK308835.1 | - | 6 | 903 | |||
AK310442.1 | - | 6 | 823 | |||
AY517557.1 | - | 6 | 855 | |||
BC012568.1 | - | 6 | 890 | 4102558 |