Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOCAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017794.4 | NP_060264.4 | 22, 23 | 2865 | ||
XM_005251494.3 | XP_005251551.2 | 20, 21 | 2849 | |||
XM_011517945.2 | XP_011516247.1 | 19, 20 | 2453 | |||
XM_017014852.1 | XP_016870341.1 | 21, 22 | 2639 | |||
XM_017014853.1 | XP_016870342.1 | 20, 21 | 2534 | |||
XM_017014854.1 | XP_016870343.1 | 20, 21 | 2534 | |||
XM_017014855.1 | XP_016870344.1 | 21, 22 | 2738 | |||
XM_017014856.1 | XP_016870345.1 | 21, 22 | 2679 | |||
XM_017014857.1 | XP_016870346.1 | 19, 20 | 2744 | |||
XM_017014858.1 | XP_016870347.1 | 19, 20 | 2744 | |||
XM_017014859.1 | XP_016870348.1 | 16, 17 | 2285 | |||
GenBank mRNA | AB058700.1 | - | 8, 9 | 971 | ||
AK055116.1 | - | 1, 2 | 173 | |||
AK056522.1 | - | 22, 23 | 2865 | |||
AY139834.1 | - | 22, 23 | 2864 |