Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAAF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017802.3 | NP_060272.3 | 7 | 1584 | ||
NR_075098.1 | 7 | 1522 | ||||
GenBank mRNA | AK056233.1 | - | 7 | 969 | ||
AK096700.1 | - | 6 | 931 | |||
AK098430.1 | - | 1 | 104 | |||
BC010850.1 | - | 2 | 311 | |||
BC047240.1 | - | 7 | 1584 | 5274510 |