Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHTF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039690.3 | NP_001034779.1 | 4 | 731 | ||
NM_001040146.3 | NP_001035236.1 | 4 | 704 | |||
NR_033227.1 | 3 | 673 | ||||
XM_011523167.1 | XP_011521469.1 | 3 | 610 | |||
XM_011523168.1 | XP_011521470.1 | 3 | 564 | |||
XM_017023370.1 | XP_016878859.1 | 4 | 726 | |||
GenBank mRNA | AF525164.1 | - | 4 | 711 | ||
AK000407.1 | - | 4 | 711 | |||
AL833902.1 | - | 1 | 290 | |||
BC018700.2 | - | 4 | 678 | 3504183 | ||
BC069059.1 | - | 3 | 594 |