Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017805.2 | NP_060275.2 | 8 | 2232 | ||
XM_011527053.2 | XP_011525355.2 | 8 | 2352 | |||
XM_011527054.2 | XP_011525356.2 | 8 | 2352 | |||
XM_017026914.1 | XP_016882403.1 | 8 | 2352 | |||
GenBank mRNA | AK000408.1 | - | 4 | 347 | ||
AY378097.1 | - | 8 | 2097 | |||
BC021860.1 | - | 4 | 362 | |||
BC028614.1 | - | 8 | 2104 | 4815880 |