Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAUS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166269.1 | NP_001159741.1 | 10 | 1327 | ||
NM_001166270.1 | NP_001159742.1 | 9 | 1184 | |||
NM_017815.2 | NP_060285.2 | 10 | 1319 | |||
GenBank mRNA | AK000431.1 | - | 10 | 1288 | ||
AK026016.1 | - | 10 | 1273 | |||
AK057779.1 | - | 8 | 1114 | |||
AY927599.1 | - | 5 | 567 | |||
BC001916.1 | - | 10 | 1316 | 2905474 | ||
BC002554.2 | - | 10 | 1250 | 3139198 | ||
BX247981.1 | - | 7 | 1125 | |||
BX247995.1 | - | 9 | 1136 | |||
BX248256.1 | - | 9 | 1151 |