Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145901.1 | NP_001139373.1 | 7 | 795 | ||
NM_017827.3 | NP_060297.1 | 6 | 789 | |||
GenBank mRNA | AB029948.1 | - | 6 | 789 | ||
AK000457.1 | - | 6 | 656 | |||
AK098522.1 | - | 6 | 367 | |||
AK293414.1 | - | 7 | 660 | |||
BC042912.1 | - | 6 | 643 | 5478759 |