Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017840.3 | NP_060310.1 | 4 | 858 | ||
GenBank mRNA | AB049642.1 | - | 4 | 742 | ||
AF183428.1 | - | 4 | 858 | |||
AF275806.1 | - | 1 | 65 | |||
AK000491.1 | - | 4 | 755 | |||
BC001040.2 | - | 4 | 724 | 2823370 | ||
BC019269.1 | - | 4 | 724 | 2823370 | ||
CA428594.1 | - | 1 | 335 | |||
CR457117.1 | - | 4 | 644 |