Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164245.1 | NP_001157717.1 | 11 | 1329 | ||
NM_001164246.1 | NP_001157718.1 | 11 | 1302 | |||
NM_017847.5 | NP_060317.3 | 12 | 1398 | |||
XM_011509660.1 | XP_011507962.1 | 12 | 1397 | |||
XM_011509661.1 | XP_011507963.1 | 12 | 1397 | |||
GenBank mRNA | AF214011.1 | - | 1 | 62 | ||
AK000171.1 | - | 2 | 185 | |||
AK000512.1 | - | 12 | 1319 | |||
AK026086.1 | - | 10 | 968 | |||
AK298107.1 | - | 11 | 1238 | |||
AY854248.1 | - | 12 | 1291 | |||
BC003397.1 | - | 12 | 1231 | 3448518 | ||
DQ323046.1 | - | 11 | 1162 | |||
DQ529299.1 | - | 10 | 1093 |