Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316943.1 | NP_001303872.1 | 3 | 845 | ||
NM_017851.5 | NP_060321.3 | 3 | 845 | |||
XM_006720589.2 | XP_006720652.1 | 3 | 886 | |||
XM_006720590.2 | XP_006720653.1 | 3 | 886 | |||
XM_006720592.2 | XP_006720655.1 | 3 | 829 | |||
XM_011521742.2 | XP_011520044.1 | 2 | 750 | |||
XM_017022386.1 | XP_016877875.1 | 3 | 829 | |||
XM_017022387.1 | XP_016877876.1 | 2 | 750 | |||
GenBank mRNA | AK058010.1 | - | 3 | 429 | ||
BC006389.1 | - | 3 | 691 | 4127509 | ||
BC031074.1 | - | 3 | 494 | 5274010 |