Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GEMIN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042479.1 | NP_001035944.1 | 5 | 930 | ||
NM_001042480.1 | NP_001035945.1 | 4 | 848 | |||
NM_017856.2 | NP_060326.1 | 5 | 1018 | |||
XM_005274555.2 | XP_005274612.1 | 4 | 3883 | |||
XM_017029617.1 | XP_016885106.1 | 4 | 1019 | |||
XM_017029618.1 | XP_016885107.1 | 4 | 1019 | |||
GenBank mRNA | AK000521.1 | - | 5 | 995 | ||
AK125062.1 | - | 4 | 2975 | |||
BC003607.2 | - | 4 | 772 | 2987964 | ||
BC014324.1 | - | 4 | 829 | 3930643 | ||
BC020785.1 | - | 4 | 825 | 4808527 | ||
BX386466.2 | - | 5 | 895 | |||
CR457241.1 | - | 2 | 675 | |||
DQ224033.1 | - | 2 | 675 |