Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017867.2 | NP_060337.2 | 6 | 748 | ||
XM_017008332.1 | XP_016863821.1 | 6 | 760 | |||
GenBank mRNA | AK000541.1 | - | 6 | 703 | ||
AL136673.1 | - | 6 | 718 | |||
BC010367.1 | - | 6 | 714 | 4334172 | ||
BM905677.1 | - | 6 | 668 |