Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THG1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317824.1 | NP_001304753.1 | 4, 5 | 579 | ||
NM_001317825.1 | NP_001304754.1 | 4, 5 | 670 | |||
NM_001317826.1 | NP_001304755.1 | 5, 6 | 642 | |||
NM_017872.4 | NP_060342.2 | 4, 5 | 674 | |||
XM_017009603.1 | XP_016865092.1 | 4, 5 | 609 | |||
GenBank mRNA | AK000553.1 | - | 4, 5 | 650 | ||
AK021663.1 | - | 4, 5 | 625 | |||
AK023355.1 | - | 3, 4 | 288 | |||
AK223119.1 | - | 4, 5 | 643 | |||
AK304590.1 | - | 4, 5 | 550 | |||
AL136669.1 | - | 4, 5 | 642 | |||
AY463216.1 | - | 4, 5 | 748 | |||
BC001523.2 | - | 4, 5 | 620 | 3010614 | ||
BC001852.2 | - | 4, 5 | 620 | 3010614 | ||
BC023521.2 | - | 4, 5 | 640 | 4126840 | ||
CR533503.1 | - | 4, 5 | 622 |