Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017882.2 | NP_060352.1 | 2, 3 | 366 | ||
GenBank mRNA | AK000568.1 | - | 2, 3 | 311 | ||
AK027604.1 | - | 2, 3 | 334 | |||
AK124013.1 | - | 2, 3 | 640 | |||
AK291175.1 | - | 2, 3 | 293 | |||
AK293155.1 | - | 2, 3 | 294 | |||
AK293197.1 | - | 2, 3 | 542 | |||
BC010849.1 | - | 2, 3 | 280 | 3878776 | ||
BC013130.1 | - | 2, 3 | 217 | 4517748 | ||
CR457244.1 | - | 2, 3 | 208 | |||
KU178671.1 | - | 2, 3 | 208 | |||
KU178672.1 | - | 2, 3 | 208 | |||
KU178674.1 | - | 2, 3 | 208 |