Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017882.2 | NP_060352.1 | 3 | 413 | ||
GenBank mRNA | AK000568.1 | - | 3 | 358 | ||
AK027604.1 | - | 3 | 381 | |||
AK124013.1 | - | 3 | 687 | |||
AK291175.1 | - | 3 | 340 | |||
AK293155.1 | - | 3 | 341 | |||
AK293197.1 | - | 3 | 589 | |||
BC010849.1 | - | 3 | 327 | 3878776 | ||
BC013130.1 | - | 3 | 264 | 4517748 | ||
CR457244.1 | - | 3 | 255 | |||
KU178671.1 | - | 3 | 255 | |||
KU178672.1 | - | 3 | 255 | |||
KU178674.1 | - | 3 | 255 |