Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017882.2 | NP_060352.1 | 4 | 623 | ||
GenBank mRNA | AK000568.1 | - | 4 | 568 | ||
AK027604.1 | - | 4 | 591 | |||
AK291175.1 | - | 4 | 550 | |||
AK293197.1 | - | 4 | 799 | |||
BC010849.1 | - | 4 | 537 | 3878776 | ||
BC013130.1 | - | 4 | 474 | 4517748 | ||
CR457244.1 | - | 4 | 465 | |||
KU178671.1 | - | 4 | 465 | |||
KU178672.1 | - | 4 | 465 | |||
KU178673.1 | - | 3 | 366 |