Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TESC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168325.1 | NP_001161797.1 | 3 | 322 | ||
NM_017899.3 | NP_060369.3 | 4 | 403 | |||
NR_031766.2 | 4 | 403 | ||||
XM_011538503.1 | XP_011536805.1 | 4 | 218 | |||
XM_017019532.1 | XP_016875021.1 | 4 | 447 | |||
XM_017019533.1 | XP_016875022.1 | 4 | 447 | |||
GenBank mRNA | AF443207.1 | - | 4 | 360 | ||
AK000614.1 | - | 4 | 446 | |||
BC015221.1 | - | 4 | 360 | 3885280 | ||
BQ434057.1 | - | 4 | 336 | |||
BQ949772.1 | - | 3 | 276 | |||
DB452909.1 | - | 4 | 403 |