Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271937.1 | NP_001258866.1 | 5 | 440 | ||
NM_017909.3 | NP_060379.2 | 6 | 958 | |||
XM_005267040.3 | XP_005267097.1 | 5 | 439 | |||
XM_017010988.1 | XP_016866477.1 | 6 | 957 | |||
GenBank mRNA | AK000634.1 | - | 6 | 901 | ||
AK026480.1 | - | 5 | 460 | |||
AK292339.1 | - | 6 | 933 | |||
BC106065.1 | - | 6 | 920 | 6165385 | ||
BC119683.1 | - | 6 | 823 | 40116343 | ||
BG547724.1 | - | 5 | 379 |