Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM118A SIRNA Assays ›
Gene Symbol:
FAM118A
Gene Name:
family with sequence similarity 118 member A
Gene Aliases:
C22orf8
Chromosome Location:
Chr.22: 45309200 - 45341955 on Build GRCh38
UniGene ID:
Hs.265018
Species:
Human
siRNA ID
HSS123713
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM118A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001104595.1
NP_001098065.1
4
897
NM_017911.2
NP_060381.2
3
570
XM_011530247.2
XP_011528549.1
3
674
XM_011530254.2
XP_011528556.1
4
593
XM_011530256.1
XP_011528558.1
3
462
XM_017028841.1
XP_016884330.1
3
459
XM_017028842.1
XP_016884331.1
3
462
XM_017028843.1
XP_016884332.1
3
459
XM_017028844.1
XP_016884333.1
3
462
XM_017028845.1
XP_016884334.1
4
659
XM_017028846.1
XP_016884335.1
3
543
XM_017028847.1
XP_016884336.1
3
1187
XM_017028848.1
XP_016884337.1
3
585
XM_017028849.1
XP_016884338.1
3
421
XM_017028850.1
XP_016884339.1
3
544
XM_017028851.1
XP_016884340.1
3
1175
XM_017028852.1
XP_016884341.1
3
581
XM_017028853.1
XP_016884342.1
3
462
XM_017028854.1
XP_016884343.1
3
544
XM_017028855.1
XP_016884344.1
4
594
XM_017028856.1
XP_016884345.1
4
594
XM_017028857.1
XP_016884346.1
3
543
XM_017028858.1
XP_016884347.1
3
544
XM_017028859.1
XP_016884348.1
3
543
XM_017028860.1
XP_016884349.1
3
544
XM_017028861.1
XP_016884350.1
3
544
GenBank mRNA
AK000642.1
-
3
540
AK125027.1
-
4
1115
BC013696.1
-
3
327
3860694
CR456403.1
-
2
296
DB153788.1
-
3
356
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-69b65fdbbd-bgr95:80/100.66.79.82:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline