Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIH1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017916.2 | NP_060386.1 | 4 | 655 | ||
GenBank mRNA | AK000650.1 | - | 4 | 607 | ||
AK304476.1 | - | 4 | 706 | |||
BC001108.2 | - | 4 | 585 | 2966990 |