Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017931.2 | NP_060401.2 | 2, 3 | 180 | ||
XM_011530260.2 | XP_011528562.1 | 2, 3 | 180 | |||
GenBank mRNA | AK000706.1 | - | 2, 3 | 136 | ||
AK300606.1 | - | 2, 3 | 160 | |||
BC018918.2 | - | 2, 3 | 121 | 4130906 | ||
DA762669.1 | - | 2, 3 | 182 |