Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017946.3 | NP_060416.1 | 3, 4 | 761 | ||
NR_046478.1 | 4, 5 | 853 | ||||
NR_046479.1 | 2, 3 | 609 | ||||
GenBank mRNA | AK000738.1 | - | 3, 4 | 613 | ||
AK309552.1 | - | 2, 3 | 470 | |||
AY358643.1 | - | 3, 4 | 641 | |||
BC005206.1 | - | 3, 4 | 644 | 4042173 | ||
DB498479.1 | - | 4, 5 | 473 |