Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTCD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017952.5 | NP_060422.4 | 23 | 1906 | ||
GenBank mRNA | AK000765.1 | - | 13 | 1077 | ||
AK315676.1 | - | 17 | 1360 | |||
AL832520.1 | - | 12 | 910 | |||
AY277598.1 | - | 11 | 874 | |||
BC001758.1 | - | 12 | 980 | |||
BC011832.2 | - | 23 | 1841 | 3939273 |