Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTCD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017952.5 | NP_060422.4 | 2, 3 | 233 | ||
GenBank mRNA | AK293962.1 | - | 2, 3 | 176 | ||
BC011832.2 | - | 2, 3 | 168 | 3939273 | ||
BI825315.1 | - | 2, 3 | 220 | |||
DA479719.1 | - | 2, 3 | 234 |