Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017956.3 | NP_060426.2 | 1 | 1157 | ||
GenBank mRNA | AK000779.1 | - | 1 | 1145 | ||
AK222727.1 | - | 1 | 1145 | |||
BC003057.2 | - | 1 | 1102 | |||
BC011713.2 | - | 1 | 1132 | 3929724 |