Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCWPW1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258008.1 | NP_001244937.1 | 11, 12 | 1436 | ||
NM_017984.4 | NP_060454.3 | 11, 12 | 1433 | |||
XM_005250479.2 | XP_005250536.1 | 11, 12 | 1870 | |||
XM_005250480.2 | XP_005250537.1 | 11, 12 | 1870 | |||
XM_006716035.3 | XP_006716098.1 | 11, 12 | 1870 | |||
XM_006716036.2 | XP_006716099.1 | 11, 12 | 1374 | |||
XM_006716037.3 | XP_006716100.1 | 11, 12 | 1867 | |||
XM_006716038.3 | XP_006716101.1 | 11, 12 | 1867 | |||
XM_006716040.3 | XP_006716103.1 | 11, 12 | 1870 | |||
XM_017012379.1 | XP_016867868.1 | 11, 12 | 1867 | |||
XM_017012380.1 | XP_016867869.1 | 11, 12 | 1371 | |||
GenBank mRNA | AK000919.1 | - | 6, 7 | 820 | ||
AK093038.1 | - | 11, 12 | 1374 | |||
AK300745.1 | - | 11, 12 | 1292 | |||
AK302108.1 | - | 10, 11 | 1170 | |||
AL136735.1 | - | 11, 12 | 1304 | |||
BC002725.1 | - | 8, 9 | 1134 | 3631266 | ||
KU178677.1 | - | 6, 7 | 696 | |||
KU178678.1 | - | 6, 7 | 699 | |||
KU178679.1 | - | 6, 7 | 696 | |||
KU178680.1 | - | 6, 7 | 699 | |||
KU178681.1 | - | 6, 7 | 696 | |||
KU178682.1 | - | 6, 7 | 696 |